Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005544157.2 | XP_005544214.1 | 2 - 3 | 549 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |