Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583087.2 | XP_005583144.1 | 3 - 4 | 911 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |