Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005584177.1 | XP_005584234.1 | 4 - 5 | 929 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |