Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590081.2 | XP_005590138.1 | 8 - 9 | 1601 | 64 | |
XM_005590082.2 | XP_005590139.1 | 7 - 8 | 1219 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |