Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLYWCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005591000.2 | XP_005591057.1 | 3 - 4 | 1084 | 72 | |
XM_005591001.2 | XP_005591058.1 | 3 - 4 | 633 | 72 | ||
XM_005591002.2 | XP_005591059.1 | 3 - 4 | 543 | 72 | ||
XM_015442626.1 | XP_015298112.1 | 3 - 4 | 540 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |