Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005545278.2 | XP_005545335.1 | 2 - 3 | 316 | 96 | |
XM_005545279.2 | XP_005545336.1 | 2 - 3 | 358 | 96 | ||
XM_005545280.2 | XP_005545337.1 | 2 - 3 | 323 | 96 | ||
XM_005545281.2 | XP_005545338.1 | 2 - 3 | 270 | 96 | ||
XM_005545282.2 | XP_005545339.1 | 2 - 3 | 267 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |