Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM270 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005549392.1 | XP_005549449.1 | 1 - 2 | 102 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |