Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4E1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005558605.2 | XP_005558662.1 | 4 - 5 | 373 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |