Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005569171.1 | XP_005569228.1 | 5 - 6 | 612 | 59 | |
XM_015429884.1 | XP_015285370.1 | 5 - 6 | 609 | 59 | ||
XM_015429885.1 | XP_015285371.1 | 4 - 5 | 483 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |