Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015440864.1 | XP_015296350.1 | 8 - 9 | 1003 | 99 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Probe spans exons |