Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102129273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005595957.2 | XP_005596014.1 | 5 - 6 | 587 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |