Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005557108.2 | XP_005557165.1 | 2 - 3 | 319 | 71 | |
XM_015451597.1 | XP_015307083.1 | 3 - 4 | 453 | 71 | ||
XM_015451598.1 | XP_015307084.1 | 2 - 3 | 234 | 71 | ||
XM_015451599.1 | XP_015307085.1 | 3 - 4 | 453 | 71 | ||
XM_015451600.1 | XP_015307086.1 | 3 - 4 | 453 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |