Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCGB1D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005577597.2 | XP_005577654.1 | 2 - 3 | 427 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |