Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASL11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015439096.1 | XP_015294582.1 | 2 - 3 | 798 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |