Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015443245.1 | XP_015298731.1 | 8 - 9 | 1162 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |