Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005555211.2 | XP_005555268.1 | 3 - 4 | 1115 | 94 | |
XM_005555212.2 | XP_005555269.1 | 3 - 4 | 1180 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |