Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPY1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005556212.2 | XP_005556269.1 | 2 - 3 | 1781 | 99 | |
XM_005556213.2 | XP_005556270.1 | 2 - 3 | 1001 | 99 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |