Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005566965.2 | XP_005567022.1 | 7 - 8 | 794 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |