Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005570931.2 | XP_005570988.1 | 4 - 5 | 1039 | 97 | |
XM_015430960.1 | XP_015286446.1 | 5 - 6 | 931 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |