Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015436176.1 | XP_015291662.1 | 4 - 5 | 578 | 57 | |
XM_015436177.1 | XP_015291663.1 | 7 - 8 | 1491 | 57 | ||
XM_015436178.1 | XP_015291664.1 | 5 - 6 | 681 | 57 | ||
XM_015436180.1 | XP_015291666.1 | 4 - 5 | 557 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |