Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005581583.2 | XP_005581640.1 | 2 - 3 | 395 | 124 | |
XM_005581584.2 | XP_005581641.1 | 3 - 4 | 510 | 124 | ||
XM_005581585.2 | XP_005581642.1 | 3 - 4 | 490 | 124 | ||
XM_015437095.1 | XP_015292581.1 | 3 - 4 | 537 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |