Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCEMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005587773.2 | XP_005587830.1 | 4 - 5 | 474 | 66 | |
XM_005587774.2 | XP_005587831.1 | 4 - 5 | 816 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |