Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005588865.2 | XP_005588922.1 | 1 - 2 | 3595 | 65 | |
XM_015441696.1 | XP_015297182.1 | 2 - 3 | 2641 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |