Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STYXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005539866.2 | XP_005539923.1 | 4 - 5 | 604 | 60 | |
XM_005539867.2 | XP_005539924.1 | 4 - 5 | 700 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |