Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005541370.2 | XP_005541427.1 | 4 - 5 | 692 | 61 | |
XM_005541371.2 | XP_005541428.1 | 4 - 5 | 678 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |