Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FILIP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005548313.2 | XP_005548370.1 | 4 - 5 | 1061 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |