Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ULBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005552121.2 | XP_005552178.1 | 2 - 3 | 155 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |