Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005588803.2 | XP_005588860.1 | 4 - 5 | 1269 | 77 | |
XM_005588805.2 | XP_005588862.1 | 4 - 5 | 1266 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |