Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005588953.2 | XP_005589010.1 | 3 - 4 | 328 | 106 | |
XM_005588954.2 | XP_005589011.1 | 3 - 4 | 814 | 106 | ||
XM_005588955.2 | XP_005589012.1 | 3 - 4 | 249 | 106 | ||
XM_015441787.1 | XP_015297273.1 | 2 - 3 | 279 | 106 | ||
GenBank mRNA | AB071098.1 | - | 2 - 3 | 236 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |