Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589542.2 | XP_005589599.1 | 7 - 8 | 1379 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |