Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590528.2 | XP_005590585.1 | 3 - 4 | 833 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |