Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCLAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005593131.2 | XP_005593188.1 | 10 - 11 | 2570 | 89 | |
XM_005593132.2 | XP_005593189.1 | 10 - 11 | 2002 | 89 | ||
XM_015443566.1 | XP_015299052.1 | 9 - 10 | 2251 | 89 | ||
XM_015443568.1 | XP_015299054.1 | 9 - 10 | 2200 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |