Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCLAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005593131.2 | XP_005593188.1 | 3 - 4 | 1225 | 109 | |
XM_005593132.2 | XP_005593189.1 | 3 - 4 | 657 | 109 | ||
XM_015443566.1 | XP_015299052.1 | 2 - 3 | 966 | 109 | ||
XM_015443568.1 | XP_015299054.1 | 2 - 3 | 942 | 109 | ||
XM_015443569.1 | XP_015299055.1 | 2 - 3 | 979 | 109 | ||
XM_015443570.1 | XP_015299056.1 | 2 - 3 | 963 | 109 | ||
XM_015443571.1 | XP_015299057.1 | 2 - 3 | 967 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Probe spans exons |