Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCLAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005593131.2 | XP_005593188.1 | 5 - 6 | 2062 | 110 | |
XM_005593132.2 | XP_005593189.1 | 5 - 6 | 1494 | 110 | ||
XM_015443566.1 | XP_015299052.1 | 4 - 5 | 1743 | 110 | ||
XM_015443568.1 | XP_015299054.1 | 4 - 5 | 1779 | 110 | ||
XM_015443569.1 | XP_015299055.1 | 4 - 5 | 1816 | 110 | ||
XM_015443570.1 | XP_015299056.1 | 4 - 5 | 1800 | 110 | ||
XM_015443571.1 | XP_015299057.1 | 4 - 5 | 1804 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |