Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005551215.2 | XP_005551272.1 | 2 - 3 | 520 | 86 | |
XM_005551216.2 | XP_005551273.1 | 2 - 3 | 520 | 86 | ||
XM_015448305.1 | XP_015303791.1 | 2 - 3 | 718 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |