Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005560395.2 | XP_005560452.2 | 1 - 2 | 651 | 86 | |
XM_015453322.1 | XP_015308808.1 | 1 - 2 | 650 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |