Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005569157.2 | XP_005569214.1 | 2 - 3 | 247 | 80 | |
XM_005569158.2 | XP_005569215.1 | 2 - 3 | 245 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |