Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005571634.2 | XP_005571691.1 | 2 - 3 | 721 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |