Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIPK1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005580401.2 | XP_005580458.1 | 3 - 4 | 1892 | 117 | |
XM_005580402.2 | XP_005580459.1 | 3 - 4 | 339 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |