Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPTE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015439049.1 | XP_015294535.1 | 16 - 17 | 1566 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |