Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPTE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015439049.1 | XP_015294535.1 | 29 - 30 | 2745 | 82 | |
GenBank mRNA | AB169152.1 | - | 19 - 20 | 1700 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |