Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005544649.2 | XP_005544706.1 | 6 - 7 | 878 | 57 | |
XM_005544650.2 | XP_005544707.1 | 6 - 7 | 875 | 57 | ||
XM_015442056.1 | XP_015297542.1 | 6 - 7 | 456 | 57 | ||
XM_015442066.1 | XP_015297552.1 | 6 - 7 | 457 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |