Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3H7orf26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005549074.2 | XP_005549131.1 | 2 - 3 | 675 | 74 |