Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUZP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005551334.1 | XP_005551391.1 | 1 - 2 | 55 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |