Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H12D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005552090.2 | XP_005552147.1 | 4 - 5 | 1473 | 86 | |
XM_005552092.2 | XP_005552149.1 | 4 - 5 | 1425 | 86 | ||
XM_015448948.1 | XP_015304434.1 | 4 - 5 | 991 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |