Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102118253 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005581145.2 | XP_005581202.1 | 1 - 2 | 279 | 91 |