Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102145326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005579328.2 | XP_005579385.1 | 6 | 1470 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |