Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283786.1 | NP_001270715.1 | 5 - 6 | 956 | 62 | |
XM_015454375.1 | XP_015309861.1 | 5 - 6 | 1431 | 62 | ||
GenBank mRNA | AB173006.1 | - | 5 - 6 | 956 | 62 | |
AF403770.1 | - | 5 - 6 | 794 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |