Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC9C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284984.1 | NP_001271913.1 | 1 - 2 | 913 | 59 | |
GenBank mRNA | AB056392.1 | - | 1 - 2 | 913 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |