Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287626.1 | NP_001274555.1 | 8 - 9 | 1134 | 102 | |
GenBank mRNA | AB168587.1 | - | 8 - 9 | 1134 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |